Z123323
Paper insert
round hole, diam. 3.0 mm
About This Item
Code UNSPSC :
12352200
Nomenclature NACRES :
SB.32
Produits recommandés
Matériaux
round hole
Technique(s)
IR spectroscopy: suitable
Diamètre
3.0 mm
Description générale
For making smaller KBr pellets. Use with the Aldrich macro-micro KBr pellet die (Z506699). Inserts also fit the Magnetic KBr pellet holder (Z112489) or Econo-Card (Z123544).