Principaux documents
Z108324
Leather gloves
size S
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
HA.03
Matériaux
brown gloves
leather
Taille
S
Stérilité
non-sterile
Caractéristiques
latex free
powder-free
reusable
Conditionnement
pack of 1 pair
Description générale
Constructed of specially treated leather that stays supple even at temperature extremes of -70 to 150°C. Remarkably durable, moisture, heat, and flame resistant. Provided with an elastic cinch for a no-slip fit.