Principaux documents
Z101397
Sigma-Aldrich
Aldrich® Kramer filter
diam. × L 12.5 mm × 110 mm
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Aldrich
Diam. × L
12.5 mm × 110 mm
Catégories apparentées
Description générale
With coarse-glass frit and metal-lock tip. In-line filter for use during double-tip needle transfer of air-sensitive solutions. (See Brown, H.C. Organic Synthesis via Boranes; Wiley-Interscience: New York, 1975; p.223.)
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC