Principaux documents
Y907340
Sigma-Aldrich
Valve
brass diaphragm, for CGA 580 right angle path, internal, right hand thread
About This Item
Code UNSPSC :
12142200
Nomenclature NACRES :
NA.12
Matériaux
brass diaphragm
Niveau de qualité
Paramètres
2640 psi max. pressure
Longueur
4.62 in.
Poids
628 g
Raccords
for CGA 580 right angle path, internal, right hand thread
Catégories apparentées
Autres remarques
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service