Principaux documents
WHA10539521
Whatman® protein saver card accessories
903 Dry rack with velcro, pack of 10 ea
Synonyme(s) :
Z699543, dry rack, protein saver card
About This Item
Code UNSPSC :
41111700
Nomenclature NACRES :
NB.79
Conditionnement
pack of 10 ea
Fabricant/nom de marque
Whatman 10539521
Whatman Article No. 28417176 (US reference)
Description générale
Whatman Dry Rak is designed to accommodate multiple collection forms at one time, properly air-drying the blood specimens in a suspended horizontal position. The Dry Rak is easily assembled and can be affixed to a wall or counter top with optional Velcro stickers.
Caractéristiques et avantages
  • The Dry Rak is easily assembled and can be affixed to a wall or Counter® top with optional Velcro stickers.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Counter is a registered trademark of Cyanamid Company
Whatman is a registered trademark of Cytiva