Principaux documents
WHA10380204
Whatman® quadrant folded cellulose fiber filter papers
Grade 41 FF, diam. (110 mm), pack of 500 ea
About This Item
Code UNSPSC :
41115727
Nomenclature NACRES :
NB.24
Matériaux
cellulose
Caractéristiques
Grade 41 FF
Nominal Ash Content: 0.007%
Conditionnement
pack of 500 ea
Fabricant/nom de marque
Cytiva
Diamètre
(110 mm)
Épaisseur
(215 μm)
Catégories apparentées
Description générale
Quadrant folded filter paper saves the user valuable time and provides ease of use when undertaking repetitive or multiple analyses.
  • Quadrant folded filter paper saves the user valuable time.
  • Quadrant folded filter paper provides ease of use when undertaking repetitive or multiple analyses.
Caractéristiques et avantages
  • Quadrant folded filter paper saves the user valuable time.
  • Quadrant folded filter paper provides ease of use when undertaking repetitive or multiple analyses.
Informations légales
Whatman is a registered trademark of Cytiva