Principaux documents
THS01239
ThalesNano CatCart® holder with SS connection tube
L × diam. 70 mm × 4 mm
Synonyme(s) :
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Conditionnement
pack of 1 ea
Fabricant/nom de marque
ThalesNano THS 01239
L × diam.
70 mm × 4 mm
Description générale
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Informations légales
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.