Principaux documents
SYNF901425G
Synthware pear-shaped Schlenk flask with stopcock
capacity 25 mL, joint: ST/NS 14/20
Synonyme(s) :
Synthware F901425G, modified Schlenk reaction flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F901425G
Capacité
25 mL
Description générale
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.
Informations légales
Synthware is a trademark of Kemtech America Inc.