Principaux documents
SYNF641425
Synthware pressure equalizing funnel with high vacuum valve
25 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonyme(s) :
Synthware F614425, dropping funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F641425
Capacité
25 mL
25 mL
Catégories apparentées
Description générale
With PTFE extended tip valve for semi-needle valve adjustment.
Informations légales
Synthware is a trademark of Kemtech America Inc.