Principaux documents
SYNF461025
Synthware storage flask with standard taper side arm
capacity 25 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonyme(s) :
Synthware F461025, round flask with side arm
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
round bottom
single-neck
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F461025
Capacité
25 mL
Taille de la vanne
0-4 mm
Description générale
Supplied with high vacuum valve with PTFE O-ring at tip.
Informations légales
Synthware is a trademark of Kemtech America Inc.