Principaux documents
SYNF311200
Synthware recovery flask
200 mL, joint: ST/NS 14/20
Synonyme(s) :
Synthware 311200, oval shape flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F311200
Capacité
200 mL
200 mL
Description générale
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.