Principaux documents
SYNF284060X
Synthware fritted gas dispersion tube with 90 degree angle
disc diam. 40 mm, porosity: extra coarse, stem O.D. × L 8 mm × 150 mm
Synonyme(s) :
Synthware F284060X, glass gas dispersion tube
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
colorless tube
glass tube
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F284060X
Diam. du disque
40 mm
Tige, D.E. × L
8 mm × 150 mm
Description générale
The 90° angle permits bubbles to be spread over a large area. Stem is 8 mm O.D. × 150 Length; XC stands for extra coarse porosity
Informations légales
Synthware is a trademark of Kemtech America Inc.