Principaux documents
SYNA862440
Synthware straight drying tube
joint: ST/NS 24/40
Synonyme(s) :
Synthware A862440, glass drying tube
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
colorless tube
glass tube
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A862440
Description générale
Synthware straight drying tube adapter with standard taper outer joint and lower inner joint of same size (24/40).
Informations légales
Synthware is a trademark of Kemtech America Inc.