Principaux documents
SYNA832440
Synthware inlet adapter with fritted disc
joint: ST/NS 24/40
Synonyme(s) :
Synthware A832440, glass inlet adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A832440
Catégories apparentées
Description générale
With an extra coarse disc to prevent solid from being introduced into reaction system during liquid transfer.
Informations légales
Synthware is a trademark of Kemtech America Inc.