Principaux documents
SYNA602924
Synthware modified anti-splash adapter
100 mL, top joint: ST/NS 29/42, bottom joint: ST/NS 24/40
Synonyme(s) :
Synthware A602924, Anti-splash adapter, splash head, bump trap
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS24/40
joint: ST/NS top29/42
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A602924
Capacité
100 mL
Catégories apparentées
Description générale
With two 4 mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask.
Informations légales
Synthware is a trademark of Kemtech America Inc.