Principaux documents
DWK986702
WHEATON® liquid scintillation vial with attached foil lined urea cap, HDPE
lips on vial transparent high-density polyethylene bottle, capacity (20 mL), screw cap
Synonyme(s) :
WHEATON® liquid scintillation vial with attached foil lined urea cap, glass, 20mL, HDPE vials, Liquid scintillation vials, Scintillation, WHEATON vials, caps attached, metal foil cork liner, vials
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
lips on vial transparent high-density polyethylene bottle
metal foil liner
white cap
Caractéristiques
with cap
Conditionnement
pkg of 500 ea
Fabricant/nom de marque
WHEATON® 986702
Diam. du flacon × Haut. × Larg.
(27 mm) (61 mm) (302 mm)
Capacité
(20 mL)
Raccords
22-400 neck
22-400
Caractéristiques et avantages
  • Converted from Type I borosilicate glass tubing, PET, or HDPE
  • Background counts are consistent and low; ultraviolet transmission is high
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.