Principaux documents
DWK73770-20125
Kimble® Disposable Screw Thread Culture Tube, 25Ml
About This Item
Code UNSPSC :
41121706
Nomenclature NACRES :
NB.17
Caractéristiques
Screw Thread Culture Tube
Conditionnement
pkg of 1 packs
Fabricant/nom de marque
Kimble® 73770-20125
Capacité
25 mL
Description générale
These tubes are suitable for tissue culture work and general bacteriological use.
Application
Bacteriology, RIA, coagulation and other laboratory procedures
Caractéristiques et avantages
  • 73760 series and 90 series items that end in ‘F′ have flat bottoms which contribute to stability
  • 73770 series items are produced with a round bottom
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Informations légales
KIMBLE is a registered trademark of DWK Life Sciences