Principaux documents
DWK45066C-24
KIMBLE® Solid Cap
black phenolic cap (with PTFE Liner), cap for 24-410
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
Matériaux
PTFE/rubber liner
black phenolic cap (with PTFE Liner)
Stérilité
non-sterile
Caractéristiques
class B
Conditionnement
case of 150 ea
Fabricant/nom de marque
Kimble® 45066C-24
Raccords
cap for 24-410
Description générale
Class B Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
Identified in ASTM Specification E982, Class B requirement
Caractéristiques et avantages
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Informations légales
KIMBLE is a registered trademark of DWK Life Sciences