Principaux documents
DWK227497-00G
Wheaton® Bod Bottle
capacity 300 mL, Un-numbered, bottle diam. × H 69 mm × 165 mm, pennyhead glass cap (stopper)
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
pennyhead glass cap (stopper)
Agence
suitable for SM 5210
Conditionnement
pkg of 24 ea
Fabricant/nom de marque
WHEATON® 227497-00G
Diam. du flacon × H
69 mm × 165 mm
Capacité
300 mL
Description générale
BOD Bottles are ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action. We recommend the 300mL capacity bottles for the five-day BOD test as referenced in Standard Methods for the Examination of Water and Wastewater, 21st Edition, 5210B, 2005.
Caractéristiques et avantages
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.
Contenu apparenté
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.