Principaux documents
DWK209126
Wheaton® Leak Resistant Bottle
capacity 60 mL, high-density polyethylene bottle, amber bottle, narrow-mouth bottle, bottle diam. × H 39 mm × 84 mm, 20-410
Synonyme(s) :
209126
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
amber bottle
high-density polyethylene bottle
narrow-mouth bottle
Conditionnement
pkg of 72 ea
Fabricant/nom de marque
WHEATON® 209126
Diam. du flacon × H
39 mm × 84 mm
Capacité
60 mL
Caractéristiques et avantages
Superior double seal closure for leak resistance
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.