DWK11-0055K
Wheaton® Microliter Snap Caps With Septa
black PTFE/silicone/PTFE cap
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.11
Matériaux
black PTFE/silicone/PTFE cap
Conditionnement
pkg of 100 ea
Fabricant/nom de marque
WHEATON® 11-0055K
Catégories apparentées
Application
Chromatography
Caractéristiques et avantages
- 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
- Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
- Approved for GC and LC (where noted).
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.