Principaux documents
DWK11-0055G
Wheaton® Microliter Snap Caps With Septa
green PTFE/silicone/PTFE cap
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.11
Matériaux
green PTFE/silicone/PTFE cap
Conditionnement
pkg of 100 ea
Fabricant/nom de marque
WHEATON® 11-0055G
Application
Chromatography
Caractéristiques et avantages
  • 11mm Snap Caps with installed septa offered in a wide variety of the most popular configurations.
  • Where noted, the Ring is installed between cap and septa for robotics employing magnetics to move vials.
  • Approved for GC and LC (where noted).
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.