Principaux documents
CLS88253424
Pyrex® bushing-type reducing adapter
female joint: ST/NS 34/45, male joint: ST/NS 24/40
Synonyme(s) :
glass adapter, glass adapters
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.43
Joint
female joint: ST/NS34/45
male joint: ST/NS24/40
Fabricant/nom de marque
Corning 8825-3424
Corning 8825
Disponibilité
not available in EU
Longueur
59 mm
Catégories apparentées
Description générale
Heavy walls for mechanical strength and heavy upper rim for good finger grip. Shorter than conventional type for more compact assemblies. Useful for attaching jointed thermometers.
Informations légales
PYREX is a registered trademark of Corning, Inc.