Principaux documents
CLS762413
Pyrex® HDPE stopper, improved form
stopper joint: 13
Synonyme(s) :
flask stopper, flask stoppers, stoppers, Corning® 7624
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.43
Joint
joint: stopper13
Fabricant/nom de marque
Corning 7624-13
Description générale
Blue and white stoppers with closed bottoms, made of linear high-density polyethylene. Improved design fully protects the neck in the event of accidental tipping.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.