Principaux documents
CLS703202L
Pyrex® VISTA round-bottom boiling flask
capacity 2,000 mL
Synonyme(s) :
boiling flask, bottom flask, round bottomed flask, Corning® 70320
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom flask
short
Joint
joint ST/NS24/40
Stérilité
non-sterile
Caractéristiques
closure type Standard taper joint
Conditionnement
case of 2
Fabricant/nom de marque
Corning 70320-2L
Capacité
2,000 mL
Taille du col
(1)
Description générale
These round bottom boiling flasks have full length outer joints, but with shorter necks. Wall thicknesses are controlled to provide balance between thermal expansion and mechanical strength for greater shock resistance.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.