Principaux documents
CLS6947500
Pyrex® resin reaction kettles
capacity 500 mL
Synonyme(s) :
pyrex flask, pyrex with lid, pyrex with lids, Corning® 6947
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Conditionnement
case of 1 ea
Fabricant/nom de marque
Corning 6947-500
Disponibilité
not available in EU
Capacité
500 mL
Description générale
Designed for viscous reaction materials. 500 and 1,000 mL kettles have interchangeable covers with four openings (24/40 joints). 2,000, 3,000 and 4,000 mL kettles have three openings with 24/40 joints and one with a 34/45 joint. Flanges of covers and bottoms are finely ground for a tight seal. Order clamp separately.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.