Principaux documents
CLS69413L
Pyrex® animal jar
O.D. × H 152 mm × 203 mm
Synonyme(s) :
Corning® 6941
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Corning 6941-3L
D.E. × H
152 mm × 203 mm
Capacité
2.8 L
Catégories apparentées
Description générale
Recommended for use as animal jars or for other purposes where resistance to hot air and steam sterilization is of importance. These jars are blown with walls approximately 4.5 mm (3/16 in.) thick. The open ends are finished with a fire-polished bead to increase mechanical strength. Do not use with heat, pressure or vacuum applications.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.