Principaux documents
CLS653402L
PYREXPLUS® heavy-wall filtering flask
capacity 2,000 mL
Synonyme(s) :
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 65340
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper9
Stérilité
non-sterile
Caractéristiques
closure type Rubber stopper
Conditionnement
pack of 1
case of 4
Fabricant/nom de marque
Corning 65340-2L
Plage de mesure du volume
600-2,000 mL
Capacité
2,000 mL
Description générale
Flask features a protective PVC coating. Autoclavable (121 °C) and resistant to thermal shock. Tubulation O.D. is 10 mm on sizes up through 1,000 mL. Tubulation is 12.5 mm O.D. on 2,000 and 4,000 mL sizes. The tubulation is not coated. Do not place over direct heat or flame. Do not heat above 121 °C moist heat or 110 °C dry heat.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREXPLUS is a registered trademark of Corning, Inc.