Principaux documents
CLS6402500
Pyrex® Squibb separatory funnel, Pyrex jointed stopper, PTFE stopcock
Synonyme(s) :
Corning® 6402
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Taille du robinet d'arrêt
4
Taille de l'obturateur
27
Fabricant/nom de marque
Corning 6402-500
Tige, D.E. × L
10 mm × 65 mm
Autres remarques
Designed so that only PTFE and borosilicate glass are in contact with liquids or gases. Two sealing mechanisms are utilized. First, the tip of the PTFE body is "threaded down" into a glass seat. Second, an adjustable gland seal within PTFE body prevents leakage through the cap. No lubrication, O-rings, or retaining clips required.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.