Principaux documents
CLS55640200
Pyrex® class A low-actinic volumetric flask, Pyrex® ST stopper
capacity 200 mL
Synonyme(s) :
flasks glass, volumetric flasks, volumetric glass
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Joint
joint stopper13
Fabricant/nom de marque
Corning 55640-200
Corning 55640
tol. measuring range
0.10 mL
Capacité
200 mL
Description générale
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of flask, which retains mechanical strength, chemical stability, and thermal resistance of Pyrex brand labware.
Informations légales
PYREX is a registered trademark of Corning, Inc.