Principaux documents
CLS55020250
Pyrex® low actinic narrow mouth Erlenmeyer flask, Pyrex® ST 27 stopper
capacity 250 mL
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flask with stopper, flasks erlenmeyer, Corning® 55020
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
narrow-mouth
Stérilité
non-sterile
Caractéristiques
closure type ST stopper
Conditionnement
case of 1
Fabricant/nom de marque
Corning 55020-250
Capacité
250 mL
Description générale
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of the flask, which retains the mechanical strength, chemical stability and thermal resistance of Pyrex brand labware.
Autres remarques
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.