Principaux documents
CLS49952L
Pyrex® wide-mouth graduated Erlenmeyer flask with screw-cap
capacity 2,000 mL
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 4995
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
polypropylene cap
screw top cap
wide-mouth
Stérilité
non-sterile
Caractéristiques
cap
closure type Screw cap
Conditionnement
case of 1
Fabricant/nom de marque
Corning 4995-2L
Capacité
2,000 mL
Description générale
Features a GL 45 size, PP, plug-seal screw-cap on a wide, 29 mm neck opening that accommodates larger volume serological pipettes. Also good for media prep and storage, or as a culture flask when optional membrane cap is used.
Autres remarques
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.