Principaux documents
CLS498010
Pyrex® narrow-mouth graduated Erlenmeyer flask
capacity 10 mL, stopper joint: 00
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks glass, Corning® 4980
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
narrow-mouth
Joint
joint: stopper00
Stérilité
non-sterile
Caractéristiques
cap: no
Conditionnement
case of 12
Fabricant/nom de marque
Corning 4980-10
Disponibilité
not available in EU
Capacité
10 mL
Description générale
  • Pyrex® narrow-mouth graduated Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
  • Flasks have durable white enamel graduations and an extra-large marking spot.
  • Features an improved tooled heavy-duty top finish.
Approximate graduations are in durable white enamel. An extra large marking space is also provided. Heavy, durable top tooling.
Autres remarques
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.