Principaux documents
CLS44602L
Pyrex® baffled trypsinizing flask, with beaded neck
capacity 1,500 mL
Synonyme(s) :
Corning® 4460
About This Item
Code UNSPSC :
41122104
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
narrow-mouth
Stérilité
non-sterile
Caractéristiques
closure type Cotton plugs
Conditionnement
case of 2
Fabricant/nom de marque
Corning 4460-2L
Capacité
1,500 mL
Description générale
Flasks are used for converting homogenous tissue samples into cell suspensions by digestion of connective proteins. Heavy beaded neck accepts cotton plugs. Four extra-deep baffles enhance vigorous agitation.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.