Principaux documents
CLS3024PACK
Pyrex® single metric scale graduated cylinder, calibrated to deliver
assortment pack
Synonyme(s) :
Corning® 3024
About This Item
Code UNSPSC :
41121805
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Corning 3024-PACK
Description générale
  • Removable PE bumperguard supplied with 25 mL through 1,000 mL sizes
The assortment pack is designed for the low volume user and contains one each of 10 mL, 25 mL, 50 mL, 100 mL, 250 mL.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.