Principaux documents
CLS2480500
Pyrex® Allihn condenser, with male and female joints
jacket L 500 mm, joint: ST/NS 24/40
Synonyme(s) :
laboratory condenser
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
Fabricant/nom de marque
Corning 2480-500
L de l'enveloppe
500 mm
Catégories apparentées
Description générale
Suitable for use in vacuum distillation. With drip-tip
Informations légales
PYREX is a registered trademark of Corning, Inc.