Principaux documents
CLS2400300
Pyrex® Liebig condenser with top and bottom joints
jacket L 300 mm, joint: ST/NS 24/40
Synonyme(s) :
Corning® 2400
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS24/40
Fabricant/nom de marque
Corning 2400-300
Disponibilité
not available in EU (Not available in EU countries)
L de l'enveloppe
300 mm
Catégories apparentées
Description générale
With drip-tip. Large-capacity water jacket cools efficiently.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.