Principaux documents
CLS1760500
Pyrex® tall form gas washing bottle, with ST stopper
capacity 500 mL
Synonyme(s) :
glass bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle
Caractéristiques
cap
Conditionnement
case of 1 ea
pack of 1 ea
Fabricant/nom de marque
Corning 1760-500
Capacité
500 mL
Diam. × H
67 mm × 300 mm
Informations légales
PYREX is a registered trademark of Corning, Inc.