Principaux documents
CLS159613L
Pyrex® graduated solution bottle
capacity 13.25 L
Synonyme(s) :
glass bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Stérilité
non-sterile
Fabricant/nom de marque
Corning 1596-13L
Capacité
13.25 L
Description générale
  • Pyrex® graduated solution bottles are best suited for sample storage and media preparation.
  • With black enamel double scale (liters and gallons) graduations in 500 mL increments.
  • Bottles can withstand both dry hot air and steam sterilization.
  • Necks are tooled to increase mechanical strength and for uniform stopper fit.
Informations légales
PYREX is a registered trademark of Corning, Inc.