Principaux documents
CAD9923B
Animal feeding needle, sterile, disposable, PTFE w/SS bite protector
size 18G, L × diam. 2 in. × 2 mm , ball
About This Item
Code UNSPSC :
42142609
Nomenclature NACRES :
NB.03
Taille
18G
Conditionnement
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
L × diam.
2 in. × 2 mm , ball
Catégories apparentées
Description générale
  • Flexible PTFE tube, covered at the hub end with type 304 stainless steel preventing tube damage due to biting during use
  • PTFE tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery
  • Plastic Luer hubs and soft silicone tips
  • Non-toxic and no-pyrogenic
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box