Principaux documents
CAD6200
1-way Luer-to-closed end adapter
FLL to closed end (316SS)
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.03
Conditionnement
pack of 5 ea
Fabricant/nom de marque
Cadence Science 6200
Catégories apparentées
Description générale
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Informations légales
Micro-Mate is a registered trademark of Cadence Science