Principaux documents
BR82122
BRAND® needle-valve stopcocks for burettes and automatic burettes with PTFE spindle
Lateral, for burette capacity 50 mL
Synonyme(s) :
Accessories, Burettes, Needle-valve stopcock, Stopcocks
About This Item
Code UNSPSC :
40141602
Nomenclature NACRES :
NB.43
Matériaux
polystyrene
Caractéristiques
for burette capacity 50 mL
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 82122
Description générale
Interchangeable PTFE valve spindle, PP cap and retention device. Valve opening 0-25.5 mm.
Please note: PTFE valve spindle operates smoothly yet is tight, even without lubrication, so there is no grease residue on the burette. The valve spindle is replaceable. Tight seal without force, a gentle turn is all it takes to close the burette valve.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG