Principaux documents
BR759242
BRAND® cap for UV-cuvette micro
green
Synonyme(s) :
Accessories, Caps, Cuvettes, UV-Cuvettes
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
SB.32
Matériaux
PE
green
polyethylene
Conditionnement
pack of 100 ea
Fabricant/nom de marque
BRAND 759242
Description générale
Round covers allow a tight closure and make it possible to store samples down to -20 °C.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG