Principaux documents
BR705854
BRAND® Transferpette® S Pipette
tip volume (500 μL), single-channel, fixed volume, mechanical, DE-M
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Caractéristiques
single-channel fixed volume
mechanical
Conditionnement
case of 1 ea
Fabricant/nom de marque
BRAND 705854
Volume de la pointe
(500 μL)
Caractéristiques et avantages
  • Efficient: one-hand operation
  • Flexible: ergonomic design for all hand sizes
  • Reliable: volume-change protection prevents accidental adjustments
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG