Principaux documents
BR705398
BRAND® Transferpette® multichannel, electronic pipette
new
8-channel, volume 50—1250 μL, AC/DC input 100 V / 240 V AC, 50—60 Hz, universal plug set
Synonyme(s) :
8-channel, Transferpette, electronic, micropipette, micropipettor, multichannel, pipet, pipette, pipettor
About This Item
Code UNSPSC :
41121503
Mode de stérilisation compatible
autoclavable compatible
Caractéristiques
8-channel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 705398
Volume
50—1250 μL
Alimentation par piles
(Nickel metal hydride 3 cells)
Description générale
Transferpette® electronic pipettes combine the globally proven properties of mechanical piston-operated air displacement pipettes by BRAND with the advantages of an electronic device.
Caractéristiques et avantages
  • The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
  • Easy calibration technique for making quick adjustments to the Transferpette® electronic, without tools.
  • Ergonomic, handy design
  • Clear and intuitive operation
  • Up to 4k pipetting cycles on a single charge
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG