Principaux documents
BR704542
BRAND® pump assembly seripettor®
size 10 mL
Synonyme(s) :
Accessories, Bottle-top Dispenser, Dispenser, seripettor®
About This Item
Code UNSPSC :
41121516
Nomenclature NACRES :
NB.01
Matériaux
polycarbonate
stainless steel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 704542
Taille
10 mL
Catégories apparentées
Description générale
The BRAND® Pump Assembly for Seripettor® features a high-quality actuating unit designed for precise liquid dispensing. Constructed from durable polycarbonate (PC), this assembly ensures longevity and reliability in laboratory applications. The stainless steel lifting spring enhances performance, providing smooth and efficient operation. This pump assembly is an essential component of the Seripettor® system, making it ideal for a variety of liquid handling tasks in research and analytical settings.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG