Principaux documents
BR65810
BRAND® desiccator
size 200
About This Item
Code UNSPSC :
41104510
Nomenclature NACRES :
NB.43
Description
lid PC, base & desiccant tray PP
lid PC, base & desiccant tray PP
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 65810
Taille
200
Catégories apparentées
Description générale
Polycarbonate lid. PP base and desiccant tray. Venting stopper with non-return valve in lid. Seal between lid and base: polychlorobutadiene rubber (CK) O-ring, compressed by vacuum. Light-weight and easy to handle.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG