Principaux documents
BR114935
BRAND® mini cooler
PC, for 12 tubes
Synonyme(s) :
Accessories, Microcentrifuge Tubes, Mini cooler, Sample Storage, Tubes
About This Item
Code UNSPSC :
41103025
Nomenclature NACRES :
NB.22
Matériaux
PC
Description
bench temp.maintained -20°C time 60 min.
Caractéristiques
for 12 tubes
Fabricant/nom de marque
BRAND 114935
Catégories apparentées
Description générale
PC. Mini coolers are designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench. Durable polycarbonate filled with non-toxic gel. Mini coolers hold twelve 0.5 ml to 2.0 ml tubes.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG