Principaux documents
BAH367150000
SP Bel-Art® Quaver Vibrating Spatula
L 7 in. (17.78 cm), brown FEP blade
Synonyme(s) :
spoon
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Matériaux
brown FEP blade
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art H36715-0000
Longueur
7 in. (17.78 cm)
Description générale
Make accurate additions of precious materials as needed. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving you control over transfer from spatula to weighing dish.
Caractéristiques et avantages
  • Available with a plain stainless steel blade or with a brown FEP coating
  • Coating complies with FDA regulations governing direct food contact applications; reduces transfer losses
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP