BAF371200125
SP Bel-Art® Spinbar® magnetic stir bar
polygon white PTFE-coated (no pivot ring), L × diam. 0.5 in. (12 mm) × 0.1875 in. (5 mm)
Synonyme(s) :
Spinbar® magnetic stir bar
About This Item
Produits recommandés
Matériaux
polygon white PTFE-coated (no pivot ring)
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F37120-0125
L × diam.
0.5 in. (12 mm) × 0.1875 in. (5 mm)
Catégories apparentées
Description générale
- Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
- Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
- Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
- Autoclave safe.
Caractéristiques et avantages
- Multi-faceted polygon shape with increased surface area creates added turbulence relative to similar sized, smooth cylindrical bars to provide thorough mixing
- Absence of a pivot ring allow the Spinbar to stay close to the bottom of the vessel enabling “scraping” of solids
- Optimal coupling with magnetic spindrive as entire length of stir bar is in close contact
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating; superior chemical compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe